Learn More
Med Vet International Rochester-Pean Forceps, 6.25 in., Straight, Von Klaus German Surgical Steel
Catalog No. 503046794
Encompass_Preferred
Rochester-Pean Forceps are used for clamping larger tissue and vessels. Straight with serrated jaws. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6794 Supplier Med Vet International Supplier No. VK1161016
Rochester-Pean Forceps are used for clamping larger tissue and vessels. Straight with serrated jaws. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.