Learn More
Med Vet International Kern Bone Holding Forceps, 9.5", with Ratchet, German Grade, Von Klaus
Catalog No. 503046617
Encompass_Preferred
9.5in. Kern bone holding forceps is a German grade surgical instruments. They feature a ratchet and a curved bottom for additional grip and control. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-6617 Supplier Med Vet International Supplier No. VK1353524
9.5in. Kern bone holding forceps is a German grade surgical instruments. They feature a ratchet and a curved bottom for additional grip and control. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.