Learn More
Med Vet International Miltex Nugent Utility Forceps 4-3/8in 45D Angled Jaws Cross-Serrated 1 2mm Wide At Tip German 18-958
Catalog No. 503071564
Encompass_Preferred
Miltex Nugent Utility Forceps 4-3/8in 45D Angled Jaws Cross-Serrated 1 2mm Wide At Tip German 18-958
This item is not returnable. View return policy
Catalog No. 50-307-1564 Supplier Med Vet International Supplier No. MIL18958
Miltex Nugent Utility Forceps 4-3/8in 45D Angled Jaws Cross-Serrated 1 2mm Wide At Tip German 18-958