Learn More
Med Vet International Miltex MeisterHand Dressing Forceps, 6in, Standard, Serrated Tips, Serrated Handles, German, MH6-10
Supplier:  Med Vet International MILMH610
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical feel of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Dressing Forceps. Dressing forceps are used when dressing wounds. They hold gauze and other dressings. They may also be used during wound debridement to remove infected or necrotic tissue or debris from the wound. They may also be used for suturing. They are made from premium-grade German stainless steel. Specifications: 6in, Standard, Serrated Tips, Serrated Handles, MH6-10
Catalog No. 50-255-9500
 Restricted Item