Learn More
Med Vet International Miltex Vantage Lorna Non-Perforating Towel Forceps, 5.25in
Supplier:  Med Vet International MILV97515
Encompass_Preferred
The Miltex Vantage V97-515 Lorna non-perforating towel forceps are made from high quality Japanese stainless steel and measure 5.25" in length. They have a finger ring handle with a ratchet lock.
Catalog No. 50-255-9419