Learn More
Med Vet International Miltex Rochester-Carmalt Forceps, 6.25in, Curved, 12/Pack, EG7-172
Supplier:  Med Vet International MILEG7172
Encompass_Preferred
The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical feel of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Rochester-Carmalt Forceps. Nicknamed the 'stars and stripes hemostat,' the Rochester-Carmalt Forceps are characterized by the longitudinal serrations that run the length of the blade with cross-hatching at the tip. These large, crushing hemostatic forceps are a choice instrument for clamping blood vessels and large tissues or ligating pedicles. They are made from premium-grade surgical stainless steel. Specifications: 6.25in, Curved, 12/Pack, EG7-172
Catalog No. 50-255-9501