Learn More
Med Vet International Miltex Rochester-Carmalt Hemostatic Forceps, 6.25in, Curved
Supplier:  Med Vet International MIL7172
Encompass_Preferred
The Miltex 7-172 Rochester-Carmalt forceps are made of OR grade German stainless steel and measure 6.25" in length. They have a finger ring handle with a ratchet lock and the tips are curved with longitudinal serrations.
Catalog No. 50-255-9511