Learn More
Med Vet International Miltex Rochester-Carmalt Hemostatic Forceps, 6.25in, Straight
Supplier:  Med Vet International MIL7170
Encompass_Preferred
The Miltex 7-170 Rochester-Carmalt forceps are made of OR grade German stainless steel and measure 6.25' in length. They have a finger ring handle with a ratchet lock and the tips are straight with longitudinal serrations.
Catalog No. 50-255-9512