Learn More
Med Vet International Miltex Rochester-Carmalt Hemostatic Forceps, 8in, Curved
Catalog No. 502559509
Encompass_Preferred
The Miltex 7-182 Rochester-Carmalt forceps are made of OR grade German stainless steel and measure 8" in length. They have a finger ring handle with a ratchet lock and the tips are curved with longitudinal serrations.
Catalog No. 50-255-9509 Supplier Med Vet International Supplier No. MIL7182
The Miltex 7-182 Rochester-Carmalt forceps are made of OR grade German stainless steel and measure 8" in length. They have a finger ring handle with a ratchet lock and the tips are curved with longitudinal serrations.