Learn More
Med Vet International Miltex Rochester-Carmalt Hemostatic Forceps, 8in, Straight
Supplier:  Med Vet International MIL7180
Encompass_Preferred
The Miltex 7-180 Rochester-Carmalt forceps are made of OR grade German stainless steel and measure 8' in length. They have a finger ring handle with a ratchet lock and the tips are straight with longitudinal serrations.
Catalog No. 50-255-9510