Learn More
Med Vet International Miltex Rochester-Pean Forceps, 12in, Curved, German, 7-148
Supplier:  Med Vet International MIL7148
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical feel of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Rochester-Pean Forceps. Rochester Pean Forceps are a hemostatic surgical forceps. This instrument is commonly referred to as a 'Big Kelly. The peans are commonly used to occlude /clamp vessels to control bleeding. They are made from premium-grade German stainless steel. Specifications: 12in, Curved, 7-148
Catalog No. 50-255-9523
 Restricted Item