Learn More
Med Vet International Miltex Plain Splinter Forceps, 3.5in, 12/Pack, EG6-300
Supplier:  Med Vet International MILEG6300
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical feel of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Plain Splinter Forceps. Plain Splinter Forceps are multi-purpose thumb forceps designed for grasping and removing small foreign objects with their fine, pointed tips. They are made from premium-grade surgical stainless steel. Specifications: 3.5in, 12/Pack, EG6-300
Catalog No. 50-255-9507