Learn More
Med Vet International Miltex Dressing Forceps, 5.5in Standard Pattern, Serrated Tips
Supplier:  Med Vet International MIL68
Encompass_Preferred
The Miltex 6-8 dressing forceps are made of OR grade German stainless steel and measure 5.5" in length. They have a non-locking thumb handle and straight, serrated tips.
Catalog No. 50-255-9537