Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spex CertiPrep™ Free Acids Analyte Mix for US EPA Method 552
SDP

Catalog No. 5522APK Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
4/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
55-22A-PK 4/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-22A-PK Supplier Spex CertiPrep™ Supplier No. 5522APK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical standards for drinking water, wastewater, and solid waste

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 552

Specifications

Packaging Bottle
CAS 79-08-3
Color Clear
Melting Point Undetermined
Boiling Point 55.2°C (131.4°F)
Solution Type Standard Solution
Type Organic Pollutant Standard
Industry Type Environmental Testing
Quantity 4/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.