Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION Pico-Tag Reagent Kit, Amino Acid Analysis, Physiologic Amino Acids

Catalog No. 50784990 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Pico-Tag Reagent Kit complements the Pico-Tag Method for amino acid analysis, providing essential reagents for the analysis of Physiologic Amino Acids. It contains PITC, TEA, and Standards A/N and B, and is designed to support a streamlined amino acid analysis and quantitation platform.

  • Reagents include PITC, TEA, and Standards A/N and B for PicoTag Amino Acid Analysis of Physiologic Amino Acids.
  • Designed for amino acid analysis.
  • Reagents undergo strict quality control testing.
  • Separation mode: Reversed phase.
  • Detection method: Optical.
  • Formulation: Liquid.
  • System type: LC.
  • Shelf life upon reconstitution: 1 month.
  • Supplied as 1 package.

Catalog No. 50-784-990 Supplier Waters Corp Supplier No. WAT010947
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Pico-Tag Reagent Kit complements the Pico-Tag Method for amino acid analysis, providing essential reagents for the analysis of Physiologic Amino Acids. It contains PITC, TEA, and Standards A/N and B, and is designed to support a streamlined amino acid analysis and quantitation platform.

  • Reagents include PITC, TEA, and Standards A/N and B for PicoTag Amino Acid Analysis of Physiologic Amino Acids.
  • Designed for amino acid analysis.
  • Reagents undergo strict quality control testing.
  • Separation mode: Reversed phase.
  • Detection method: Optical.
  • Formulation: Liquid.
  • System type: LC.
  • Shelf life upon reconstitution: 1 month.
  • Supplied as 1 package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.