Learn More
Drg International Inc DRG Free β-HCG ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA4718R
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of free beta subunit of human chorionic gonadotropin (free Beta-hCG) in serum and EDTA plasma. This product is for Research Use Only. The DRG Free Beta-HCG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal [mouse] antibody directed towards a unique antigenic site on a Free Beta-hCG molecule. An aliquot sample containing endogenous Free Beta-hCG is incubated in the coated well with enzyme conjugate, which is an anti-Beta-HCG antibody [rabbit] conjugated with horseradish peroxidase.
Catalog No. 50-194-7618