Learn More
POLLARDWATER COM Free Chlorine Test Strips 0-6 ppm 30/pk
Catalog No. NC1725202
Encompass
Inexpensive and easy-to-use. Fast and accurate results. Perfect for quick chlorine checks in water, wastewater, and dichlorination applications. Easiest, most cost effective method for testing chlorine levels in water but may not meet requirements for reporting purposes. Range: 0 to 6 ppm. Steps: 0, 0.05, 0.1, 0.2, 0.4, 0.6, 0.8, 1.2, 1.5, 2.0, 2.6, 4.0, 6.0 ppm.
Catalog No. NC1725202 Supplier POLLARDWATER COM Supplier No. I481126
Inexpensive and easy-to-use. Fast and accurate results. Perfect for quick chlorine checks in water, wastewater, and dichlorination applications. Easiest, most cost effective method for testing chlorine levels in water but may not meet requirements for reporting purposes. Range: 0 to 6 ppm. Steps: 0, 0.05, 0.1, 0.2, 0.4, 0.6, 0.8, 1.2, 1.5, 2.0, 2.6, 4.0, 6.0 ppm.