Learn More
Fujifilm Healthcare Americas Corp Free Cholesterol E
Catalog No. NC9506034
Encompass_Preferred
The Free Cholesterol E kit is an in vitro assay utilizing an enzymatic method for the determination of Free Cholesterol in serum. Package size: Kit includes: Buffer Solution- 2 x 75 mL, Color Reagent- 2 x for 75 mL, Standard Solution- 1 x 10 mL bottles. This product is for Research Use Only, not for use in diagnostic procedures. Contact supplier for additional information at www.ivd.fujimed.com.
Catalog No. NC9506034 Supplier Fujifilm Healthcare Americas Corp Supplier No. 99302501
The Free Cholesterol E kit is an in vitro assay utilizing an enzymatic method for the determination of Free Cholesterol in serum. Package size: Kit includes: Buffer Solution- 2 x 75 mL, Color Reagent- 2 x for 75 mL, Standard Solution- 1 x 10 mL bottles. This product is for Research Use Only, not for use in diagnostic procedures. Contact supplier for additional information at www.ivd.fujimed.com.