Learn More
Free Cyanide Standard, Potassium Cyanide, 1000 μg/mL, 125 mL
Specifications
Specifications
| CAS Max % | 99.64, 0.1, 0.25 |
| Analyte or Component Names | Water,sodium hydroxide,potassium cyanide |
| Element | Sodium Hydroxide, Potassium Cyanide |
| Solvent or Matrix | Water/Dilute Sodium Hydroxide |
| Quantity | 125 mL |
| Concentration or Composition (by Analyte or Components) | 1000μg/mL |
| Chemical Name or Material | Free Cyanide Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.