Learn More
Biotium Free Dyes In Vivo Imaging Reactive CF Dyes Other Dyes Biotinylation Reagents CF450 1/EA
Catalog No. 5000501464
Encompass_Preferred
CF Dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability CF Dye free acids are control dyes for corresponding CF Dye conjugates CF450 dye has excitation/emission at 448/533nm
Catalog No. 50-005-01464 Supplier Biotium Supplier No. 98004
Add to Cart
Add to Cart
CF Dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability CF Dye free acids are control dyes for corresponding CF Dye conjugates CF450 dye has excitation/emission at 448/533nm