Learn More
Biotium Free Dyes In Vivo Imaging Reactive CF Dyes Other Dyes Biotinylation Reagents CF640 1/EA
Catalog No. 5000501491
Encompass_Preferred
CF Dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability CF Dye free acids are control dyes for corresponding CF Dye conjugates CF640R dye has excitation/emission at 642/663 nm
Catalog No. 50-005-01491 Supplier Biotium Supplier No. 92063
Add to Cart
Add to Cart
CF Dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness and photostability CF Dye free acids are control dyes for corresponding CF Dye conjugates CF640R dye has excitation/emission at 642/663 nm