Learn More
Alpco Ltd Free Estriol ELISA
SDP
Supplier:  Alpco Ltd 20FE3HUE01
Encompass_Preferred
The Free Estriol ELISA is an enzyme immunoassay for the quantitative in vitro diagnostic measurement of free estriol (unconjugated estriol) in serum during the second half of pregnancy. FDA Registered. For In Vitro Diagnostic Use.
Catalog No. 50-751-3517