Learn More
Drg International Inc Free Estriol Elisa
Supplier:  Drg International Inc EIA1612
Encompass_Preferred
An enzyme immunoassay for the quantitative measurement of free estriol (unconjugated estriol) in serum during the second half of pregnancy. Estriol (E3) is the major estrogen formed by the fetoplacental unit during pregnancy. The DRG Free Estriol ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal [rabbit] antibody directed towards an antigenic site on the Estriol molecule.
Catalog No. 50-194-7538