Learn More
Cell Biolabs Inc Free Fatty Acid Assay Kit (Colorimetric), 100 assays
Supplier:  Cell Biolabs Inc STA618
Encompass_Preferred
Free fatty acids (FFA) are formed from the hydrolyisis of triglyceride ester bonds and serve as a ready energy source by binding to albumin in the bloodstream. Measurement of FFA has become an important part of monitoring various disease states including obesity, diabetes, and cancer. Cell Biolabs' Free Fatty Acid Assay Kit quantifies the FFA concentration (non-esterified) in various samples using a 96-well colorimetric plate format. It can detect FFA levels as low as 15 µM in plasma or serum samples. Each kit provides sufficient reagents for 100 assays including standards and unknown samples.
Catalog No. 50-673-657