Learn More
Waters Corp Free Fatty Acid HP Column, 4 µm, 3.9 mm x 150 mm, 1/pk
Catalog No. 50785045 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Free Fatty Acid HP column uses a phenyl-bonded packing to separate free fatty acids based on carbon chain length and degree of saturation.
Catalog No. 50-785-045 Supplier Waters Corp Supplier No. WAT011690
The Free Fatty Acid HP column uses a phenyl-bonded packing to separate free fatty acids based on carbon chain length and degree of saturation.