Learn More
Cayman Chemical Free Faty ACD FluorometrIc 96
SDP
Catalog No. 502895272
Encompass_Preferred
The measurement of FFA can be useful in determining metabolic status. Cayman’s Free Fatty Acid (FFA) Assay can be used for measuring free fatty acids in plasma, serum, and urine. The FFA Assay utilizes a coupled enzymatic reaction that results in generation of the highly fluorescent product resorufin.
Catalog No. 50-289-5272 Supplier Cayman Chemical Supplier No. 70031096 WELL
The measurement of FFA can be useful in determining metabolic status. Cayman’s Free Fatty Acid (FFA) Assay can be used for measuring free fatty acids in plasma, serum, and urine. The FFA Assay utilizes a coupled enzymatic reaction that results in generation of the highly fluorescent product resorufin.