Learn More
Sigma Aldrich Fine Chemicals Biosciences Free Glycerol Reagent used for quantitative enzymatic determination of glycerol, 9002-93-1
Catalog No. 501657281
Encompass
Free Glycerol Reagent used for quantitative enzymatic determination of glycerol
Catalog No. 50-165-7281 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F642840ML
Free Glycerol Reagent used for quantitative enzymatic determination of glycerol