Learn More
Eagle Biosciences Inc Prostatic Specific Antigen Free (fPSA) ELISA
SDP
Supplier:  Eagle Biosciences Inc PSF31K01
Encompass
The Eagle Biosciences Free Prostate Specific Antigen (fPSA) ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the direct quantitative determination of Free Prostate Specific Antigen in human serum. The Eagle Biosciences Free Prostate Specific Antigen (fPSA) ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.
Catalog No. 50-187-8193