Learn More
PRODUCTION BASICS FREE STANDING ADJUSTABLE FOOT REST 12"D x 20"W
Catalog No. NC1058436
Encompass_Preferred
Use the Free Standing Footrest with any station you order from Production Basics, or for your existing workstations. This footrest has an 80 degree angled foot platform with height adjustability from 1.5" to 15".
Catalog No. NC1058436 Supplier PRODUCTION BASICS Supplier No. 8659
Use the Free Standing Footrest with any station you order from Production Basics, or for your existing workstations. This footrest has an 80 degree angled foot platform with height adjustability from 1.5" to 15".