Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Free Standing Eyewash and Shower Station

Catalog No. 06000440 Shop All Fisherbrand Products
Change view
Click to view available options
For Use With (Application):
ADA Safety Shower
Safety Shower
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
06-000-440 ADA Safety Shower
06-000-439 Safety Shower
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-000-440
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Free Standing Eyewash and Shower Station includes a drench shower and eye wash fixture.

  • Drench shower is made out of ABS plastic
  • Ball valve operated by rigid pull-rod and easy grip handle
  • Eye wash fixture with stainless steel bowl, four gently spray heads and a Push to Operate ball valve
  • Eye wash and shower operate independently
  • Overall height 80 in.

Specifications

Certifications/Compliance ADA Compliant
Color Orange
Flow Rate 20 gpm
For Use With (Application) ADA Safety Shower
Includes Free-standing Drench Shower and Eyewash
Material ABS Plastic
Mounting Floor
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.