Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ Free T4 (Thyroxine) Coated Tube RIA Kit

Catalog No. MP06B257215 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP06B257215 500 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP06B257215 Supplier MP Biomedical™ Supplier No. 06B257215
Add to Cart
Add to Cart

Quantitative Solid Phase Radioimmunoassay

The Free T4 (Thyroxine) Coated Tube RIA Kit is intended for the measurement of Free T4 in a sample.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Kit and Assay
Sample Type Plasma, Serum
Target Species Human
For Use With (Application) Endocrinology
Host Species Human
Quantity 500 Tubes
Regulatory Status FDA Cleared (K823264/A)
Sample Volume 50 μL
Storage Requirements 2°C to 8°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.