Learn More
Crystal Chem Inc Free Thyroxine (fT4) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80984
Encompass
The Free Thyroxine ELISA is a highly sensitive assay that is used to quantify levels of free human thyroxine in serum. Highlights: Kits use sample size of only 25 µL. High sensitivity of 1 pg/mL. Dynamic range of 2 - 95 pg/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7885