Learn More
Hach Company Freechlor F Reagent Solution, 50 mL
Supplier:  Hach Company 2964926
Encompass
Used to determine free chlorine in the presence of manganese, chloramines and other interfering oxidants. Hach Method 10241 Requires Monochlor F Reagent, PN 2802299. 50 mL bottle. Approximately 100 tests.
Catalog No. 50-199-6957