Learn More
MSC FREEDOM RATCHET HARD HAT
Catalog No. 5000782730
Encompass
The V-Gard Slotted Protective Hat, part of the American Freedom Series, features an American Flag with 2 Eagles design. This durable head protection is recognized for its comfort and quality, identified by its distinctive 'V' design.
  • Features Fas-Trac III Suspension
  • Includes slots for improved impact resistance
  • Designed for personal comfort
  • Available with patriotic designs
  • Constructed from polyethylene
  • Non-vented for full protection
  • Meets ANSI/ISEA Z89.1-2014 and CAN/CSA Z94.1-2015 standards
Catalog No. 50-007-82730 Supplier MSC Supplier No. 72146152
The V-Gard Slotted Protective Hat, part of the American Freedom Series, features an American Flag with 2 Eagles design. This durable head protection is recognized for its comfort and quality, identified by its distinctive 'V' design.
  • Features Fas-Trac III Suspension
  • Includes slots for improved impact resistance
  • Designed for personal comfort
  • Available with patriotic designs
  • Constructed from polyethylene
  • Non-vented for full protection
  • Meets ANSI/ISEA Z89.1-2014 and CAN/CSA Z94.1-2015 standards