Learn More
ABBOTT DIABETES CARE INC Blood Glucose Meter FreeStyle Lite 5 Second Results, 4/case
Catalog No. NC1783604
Encompass
Stores Results 7 , 14 and 30 Day Averaging No Coding Required. Blood Sample Volume 0.3 Microliter.
This item is not returnable. View return policy
Catalog No. NC1783604 Supplier ABBOTT DIABETES CARE INC Supplier No. 7080470
Stores Results 7 , 14 and 30 Day Averaging No Coding Required. Blood Sample Volume 0.3 Microliter.