Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ FreeStyle™ MAX Reagent

Catalog No. 16447750
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
15 mL
10 x 15 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-447-750 10 x 15 mL
16-447-100 1 mL
16-447-500 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 16-447-750 Supplier Gibco™ Supplier No. 16447750
Only null left
Add to Cart
Add to Cart
Request a Quote

Animal origin-free formulation for transfecting plasmid DNA into eukaryotic cells that can easily be scaled up to produce large amounts of recombinant proteins

  • Allows highest expression levels and transfection rates with lowest cytotoxicity in bio-production applications
  • Formulated for use with:
  • FreeStyle 293-F Cells (suspension human embryonal kidney cells, Cat. No. R790-07) in serum-free FreeStyle 293 Expression Medium (Cat. No. 12338-018)
  • FreeStyle CHO-S Cells (suspension Chinese Hamster Ovary cells, Cat. No. R800-07) in serum-free FreeStyle CHO Expression Medium (Cat. No. 12651-014)
  • DG44 Cells (DHFR– suspension CHO cells) in CD DG44 Medium with 8 mM L-glutamine and 18 mL/L 10% Pluronic F-68 (Cat. No. A11000-01)
  • FreeStyle™ MAX Reagent is a proprietary, animal origin–free formulation for transfecting plasmid DNA into eukaryotic cells that can easily be scaled up to produce large amounts of recombinant proteins
  • Intended for use with the FreeStyle™ MAX 293 Expression System (Cat. No. K9000-10) and the FreeStyle™ MAX CHO Expression System (Cat. No. K9000-20).
TRUSTED_SUSTAINABILITY

Specifications

Classification Animal Origin-Free
Content And Storage Each kit contains 10 units. Each unit contains one vial of reagent. Store at +4°C. FreeStyle™ MAX is guaranteed stable for 6 months when stored properly.
Cell Type Established Cell Lines
Format Tube
Sample Type Plasmid DNA
Transfection Technique Lipid-based Transfection
For Use With (Application) Protein Expression
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line Freestyle
Product Type Transfection Reagent
Quantity 10 x 15 mL
Serum Compatible Yes
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.