Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lux Blox™ Freestyle Set
SCI_ED

Catalog No. S81703
Change view
Click to view available options
Grade:
Pre-K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S81703 Pre-K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81703 Supplier Lux Blox™ Supplier No. LUX166
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The perfect way for children to become immersed in the world of Lux Blox

  • One-of-a-kind hinged pieces snap together to allow children o build anything their imaginations can dream up
  • Durable building block pieces connect on all 4 sides to allow for more flexible builds, creating toys where any part can move
  • Set includes instructions for multiple builds including a Dragon, Sword, Bug Buggy, and Race Car
  • Supervised use is recommended for young children

Specifications

Color Neon Orange, Teal Blue, Black, and White
Grade Pre-K to 12
Age 6
Includes 116 Pc., Building Instructions, and Various Accessories
Quantity 1 Kit
Product Type Freestyle Set
Description Contains Enough Pieces to Construct One Item at a Time
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.