Learn More
MSC Nu-Calgon Freez-Kontr'l, Propylene Glycol, Inhibited, Corrosion Protection
Catalog No. 502840057
Encompass
Freez-Kontr'l is a formulation of propylene glycol, dipotassium phosphate corrosion inhibitor, and de-ionized water, colored blue for leak detection. It has an operating temperature range from below -60°F to 230°F, providing freeze protection to below -60°F and burst protection to below -100°F. It is nearly odorless and is the product of choice when contact with potable water is possible, as its ingredients are Generally Recognized As Safe (GRAS).
  • Optimum freeze or burst protection.
  • Fully inhibited to prevent corrosion.
  • Non-flammable.
  • Uses renewable resourced propylene glycol.
  • Low in acute oral toxicity.
  • Colored blue for easy leak detection.
Catalog No. 50-284-0057 Supplier MSC Supplier No. 88514831
Freez-Kontr'l is a formulation of propylene glycol, dipotassium phosphate corrosion inhibitor, and de-ionized water, colored blue for leak detection. It has an operating temperature range from below -60°F to 230°F, providing freeze protection to below -60°F and burst protection to below -100°F. It is nearly odorless and is the product of choice when contact with potable water is possible, as its ingredients are Generally Recognized As Safe (GRAS).
  • Optimum freeze or burst protection.
  • Fully inhibited to prevent corrosion.
  • Non-flammable.
  • Uses renewable resourced propylene glycol.
  • Low in acute oral toxicity.
  • Colored blue for easy leak detection.