Learn More
Chemglass Life Sciences FreezeCell, Hexagon Shape, Holds (12) 1.0-2.0mL Cryogenic Vials or 1.5-2.0mL Centrifuge Tubes
SDP
Catalog No. 501979512
Encompass
FreezeCell, Hexagon Shape, Holds (12) 1.0-2.0mL Cryogenic Vials or 1.5-2.0mL Centrifuge Tubes. The FreezeCell™ can be used with a variety of cell types, including stem cells, primary cells, cell lines and yeast. The FreezeCell™ doesn’t require any additives, all you need to do is place the FreezeCell™ into an ultra-low freezer, and the cooling will begin at -1⁰C per minute. It is a very good device for all protocols of cell preservation. This product is symmetrically designed with a combination of foam and alloys with no additives or emission characteristics. These products are environmentally friendly. No Need to replace any parts, and will serve you for years.
Catalog No. 50-197-9512 Supplier Chemglass Life Sciences Supplier No. CLS3729H01
FreezeCell, Hexagon Shape, Holds (12) 1.0-2.0mL Cryogenic Vials or 1.5-2.0mL Centrifuge Tubes. The FreezeCell™ can be used with a variety of cell types, including stem cells, primary cells, cell lines and yeast. The FreezeCell™ doesn’t require any additives, all you need to do is place the FreezeCell™ into an ultra-low freezer, and the cooling will begin at -1⁰C per minute. It is a very good device for all protocols of cell preservation. This product is symmetrically designed with a combination of foam and alloys with no additives or emission characteristics. These products are environmentally friendly. No Need to replace any parts, and will serve you for years.