Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Freeze Dried E. Coli JM101 Bacteria Culture
SCI_ED

Catalog No. S37738 Shop All Aldon Products
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S37738 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S37738 Supplier Innovating Science™ Supplier No. IS3907
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Freeze dried into a lyophilized pellet packaged in a sterile plastic vial.

  • Includes one tube of rehydrating broth and one tube agar slant, sterile swab and instructions

Specifications

Color White
Grade 7 to 14
Age 13 to 20
Includes One tube of rehydrating broth and one tube agar slant, sterile swab and instructions
Quantity 1
Class Size 1 Class
Product Type Freeze Dried E. Coli JM101 Bacteria Culture
Description Format: Lyophilized

DOT: Non regulated

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.