Learn More
Bellco Glass, Inc. Freeze-Dry Ampoule, 1mL Tubular
SDP
Catalog No. 501945594
Encompass_Preferred
Type 1 class a Borosilicate glass ideal for freeze drying or cryogenic storage. Tubular firing off constriction on the neck. Rigidly controller bottom to withstand pressure at low temperature.. MADE IN THE USA Tubular round bottom Length: 127mm Neck od: 7mm Capacity: 1ml
Catalog No. 50-194-5594 Supplier Bellco Glass, Inc. Supplier No. 402500001
Type 1 class a Borosilicate glass ideal for freeze drying or cryogenic storage. Tubular firing off constriction on the neck. Rigidly controller bottom to withstand pressure at low temperature.. MADE IN THE USA Tubular round bottom Length: 127mm Neck od: 7mm Capacity: 1ml