Learn More
Bellco Glass, Inc. Freeze-Dry Ampoule,1mL Flat Bottom - Constriction Neck
SDP
Catalog No. 501945590
Encompass_Preferred
Type 1 Class A Borosilicate Glass for freeze drying or cryogenic storage. The neck has pre-drawn constriction to facilitate sealing.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassFlat BottomConstriction NeckNeck OD : 7mmBottom OD: 10mmCapacity: 1mLLength 80mmCase Quantity: 72
Catalog No. 50-194-5590 Supplier Bellco Glass, Inc. Supplier No. 401900001
Type 1 Class A Borosilicate Glass for freeze drying or cryogenic storage. The neck has pre-drawn constriction to facilitate sealing.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassFlat BottomConstriction NeckNeck OD : 7mmBottom OD: 10mmCapacity: 1mLLength 80mmCase Quantity: 72