Learn More
Bellco Glass, Inc. Freeze Dry Ampoule,1mL Constriction on neck
SDP
Catalog No. 501945591
Encompass_Preferred
Type 1 class a Borosilicate glass for freeze drying or cryogenic storage. The neck has pre-drawn constriction to facilitate sealing.. MADE IN THE USA Flat bottom Constriction neck Neck od : 8mm Bottom od: 11mm Capacity: 1ml Length: 145mm
Catalog No. 50-194-5591 Supplier Bellco Glass, Inc. Supplier No. 402400001
Type 1 class a Borosilicate glass for freeze drying or cryogenic storage. The neck has pre-drawn constriction to facilitate sealing.. MADE IN THE USA Flat bottom Constriction neck Neck od : 8mm Bottom od: 11mm Capacity: 1ml Length: 145mm