Learn More
Bellco Glass, Inc. Freeze-Dry Ampoule, 2mL Tubular
SDP
Catalog No. 501945595
Encompass_Preferred
Type 1 class a Borosilicate glass ideal for freeze drying or cryogenic storage. Tubular firing off constriction on the neck. Rigidly controller bottom to withstand pressure at low temperature.. MADE IN THE USA Tubular round bottom Length:145mm Neck od: 8mm Capacity: 2ml
Catalog No. 50-194-5595 Supplier Bellco Glass, Inc. Supplier No. 402500002
Type 1 class a Borosilicate glass ideal for freeze drying or cryogenic storage. Tubular firing off constriction on the neck. Rigidly controller bottom to withstand pressure at low temperature.. MADE IN THE USA Tubular round bottom Length:145mm Neck od: 8mm Capacity: 2ml