Learn More
Bellco Glass, Inc. Freeze-Dry Ampoule,2mL Constriction on neck
SDP
Catalog No. 501945592
Encompass_Preferred
Type 1 class a Borosilicate glass for freeze drying or cryogenic storage. The neck has pre-drawn constriction to facilitate sealing.. MADE IN THE USA Flat bottom Constriction neck Neck od: 9mm Bottom od: 12mm Capacity: 2ml Length: 145mm
Catalog No. 50-194-5592 Supplier Bellco Glass, Inc. Supplier No. 402400002
Type 1 class a Borosilicate glass for freeze drying or cryogenic storage. The neck has pre-drawn constriction to facilitate sealing.. MADE IN THE USA Flat bottom Constriction neck Neck od: 9mm Bottom od: 12mm Capacity: 2ml Length: 145mm