Learn More
Bellco Glass, Inc. Freeze-Dry Ampoule,3mL Tubular
SDP
Catalog No. 501945596
Encompass_Preferred
Type 1 class a Borosilicate glass ideal for freeze drying or cryogenic storage. Tubular firing off constriction on the neck. Rigidly controller bottom to withstand pressure at low temperature.. MADE IN THE USA Tubular round bottom Length: 145mm Neck od: 8mm Capacity: 3ml
Catalog No. 50-194-5596 Supplier Bellco Glass, Inc. Supplier No. 402500003
Type 1 class a Borosilicate glass ideal for freeze drying or cryogenic storage. Tubular firing off constriction on the neck. Rigidly controller bottom to withstand pressure at low temperature.. MADE IN THE USA Tubular round bottom Length: 145mm Neck od: 8mm Capacity: 3ml