Learn More
Bellco Glass, Inc. Freeze-Dry Ampoule,4mL Tubular
SDP
Catalog No. 501945597
Encompass_Preferred
Type 1 Class A Borosilicate Glass ideal for freeze drying or cryogenic storage. Tubular firing off constriction on the neck. Rigidly controller bottom to withstand pressure at low temperature.. MADE IN THE USA Tubular round bottom Length: 170mm Neck od: 8mm Capacity: 4ml
Catalog No. 50-194-5597 Supplier Bellco Glass, Inc. Supplier No. 402500004
Type 1 Class A Borosilicate Glass ideal for freeze drying or cryogenic storage. Tubular firing off constriction on the neck. Rigidly controller bottom to withstand pressure at low temperature.. MADE IN THE USA Tubular round bottom Length: 170mm Neck od: 8mm Capacity: 4ml