Learn More
Bellco Glass, Inc. Freeze-Dry Ampoule.5mL Constriction on neck
SDP
Catalog No. 501945593
Encompass_Preferred
Type 1 class a Borosilicate glass for freeze drying or cryogenic storage. The neck has pre-drawn constriction to facilitate sealing.. MADE IN THE USA Flat bottom Constriction neck Neck od: 6mm Bottom od: 4mm Capacity: 5ml Length: 105mm
Catalog No. 50-194-5593 Supplier Bellco Glass, Inc. Supplier No. 402450000
Type 1 class a Borosilicate glass for freeze drying or cryogenic storage. The neck has pre-drawn constriction to facilitate sealing.. MADE IN THE USA Flat bottom Constriction neck Neck od: 6mm Bottom od: 4mm Capacity: 5ml Length: 105mm