Learn More
Electron Microscopy Sciences Embra Freeze Fracture Grid 75FF, Copper
SDP
Catalog No. 5026072
Encompass_Preferred
Embra electroformed grids are 3.05 mm in diameter and 16 um thick.
Type: 75FF
Material: Copper
Catalog No. 50-260-72 Supplier Electron Microscopy Sciences Supplier No. E75FFCU
Embra electroformed grids are 3.05 mm in diameter and 16 um thick.
Type: 75FF
Material: Copper