Learn More
Electron Microscopy Sciences Embra Freeze Fracture Grid 75FF, Nickel
SDP
Catalog No. 5026071
Encompass_Preferred
Embra electroformed grids are 3.05 mm in diameter and 16 um thick.
Type: 75FF
Material: Nickel
Catalog No. 50-260-71 Supplier Electron Microscopy Sciences Supplier No. E75FFNI
Embra electroformed grids are 3.05 mm in diameter and 16 um thick.
Type: 75FF
Material: Nickel